Learn More
Invitrogen™ Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31452 has been successfully used in Western blot, and ICC applications. Antibody Specificity: This antibody reacts with the heavy chains on mouse IgG and with light chains on most mouse immunoglobulins based on immunoelectrophoresis. No antibody was detected against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 1.0 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.
Specifications
Specifications
| Antigen | Mouse IgG (H+L) Cross-Adsorbed |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 15mg/mL BSA and no preservative; pH 7.6 |
| Host Species | Rabbit |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |