missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 568
Description
Product will be shipped at Room Temperature.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Mouse IgG (H+L) Cross-Adsorbed |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Alexa Fluor 568 |
| Formulation | PBS with 5mM sodium azide; pH 7.5 |
| Host Species | Rabbit |
| Immunogen | Gamma Immunoglobins Heavy and Light chains. |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |