missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Mouse, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP160FMI
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP160FMI 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP160FMI Supplier MilliporeSigma Supplier No. AP160F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Rabbit anti-Mouse IgG Antibody, FITC conjugate detects level of Mouse IgG and has been published and validated for use in immunofluoresence.

Specifications

Antigen Rabbit anti-Mouse IgG
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Host Species Rabbit
Quantity 1.5 mg
Primary or Secondary Secondary
Target Species Mouse

For Research Use Only