missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Mouse, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP160AMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP160AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP160AMI Supplier MilliporeSigma Supplier No. AP160A

Rabbit Polyclonal Antibody

Rabbit anti-Mouse IgG Antibody, Alkaline Phosphatase conjugate detects level of Mouse IgG and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Rabbit anti-Mouse IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Host Species Rabbit
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse

For Research Use Only