missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Human IgM Recombinant Secondary Antibody, Biotin

Catalog No. PISA510247
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PISA510247 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PISA510247 Supplier Invitrogen™ Supplier No. SA510247

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Secondary Antibody

Specificity: reacts to the heavy chain of human IgM. No cross reactivity with human IgG, IgA, IgD, or IgE. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human IgM
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RM121
Concentration 1.0 mg/mL
Conjugate Biotin
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Host Species Rabbit
Immunogen Human IgM.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -20°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less