missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Human, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP101PMI
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP101PMI 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP101PMI Supplier MilliporeSigma Supplier No. AP101P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Detect Human IgG using this Rabbit anti-Human IgG Antibody, HRP conjugate validated for use in ELISA and western blotting.

Specifications

Antigen Rabbit anti-Human IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Host Species Rabbit
Quantity 1.5 mL
Primary or Secondary Secondary
Target Species Human

For Research Use Only