missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Human IgG Heavy Chain Secondary Antibody [Janelia Fluor 669], R&D Systems™

Catalog No. NB785900
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB785900 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB785900 Supplier Bio-Techne Supplier No. NBP222138JF669

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot

Specifications

Antigen IgG Heavy Chain
Applications Western Blot, ELISA, Immunohistochemistry, Dot Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Rabbit
Immunogen Human IgG gamma heavy chain
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less