Learn More
Invitrogen™ Rabbit anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 550
Rabbit Polyclonal Secondary Antibody
Supplier:  Invitrogen™ SA510111
Catalog No. PISA510111
Only null left
Explore available promotions
Description
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to mouse and rat.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Human IgG (H+L) Cross-Adsorbed
Polyclonal
DyLight 550
Rabbit
Antigen affinity chromatography
RUO
Human
Secondary Antibody
IgG
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Human IgG-heavy and light chain.
500 μg
Secondary
4°C
Liquid
Product Suggestions
Product Suggestions