missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Rabbit anti-Human IgD, Clone: RM123, Secondary Antibody,

Catalog No. 041659MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
041659MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 041659MI Supplier MilliporeSigma™ Supplier No. 041659

Rabbit Monoclonal Antibody

Specifically detects Human IgD clone: RM123 in Human samples, and it is validated for ELISA

Specifications

Antigen Human IgD
Applications ELISA
Classification Monoclonal
Clone RM123
Formulation Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Host Species Rabbit
Immunogen Human IgD
Purification Method Protein A Purfied
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary and Control Antibodies
Primary or Secondary Secondary
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only