missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgA Heavy Chain Secondary Antibody [Janelia Fluor 669], R&D Systems™
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
Specifications
| Antigen | IgA Heavy Chain |
| Applications | Western Blot, ELISA, Immunohistochemistry, Dot Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Human IgA alpha heavy chain |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |