missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Goat, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP106RMI
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP106RMI 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP106RMI Supplier MilliporeSigma Supplier No. AP106R

Rabbit Polyclonal Antibody

This Rabbit anti-Goat IgG Antibody, Rhodamine conjugate is validated for use in IF for the detection of Goat IgG.

Specifications

Antigen Rabbit anti-Goat IgG
Applications Immunofluorescence
Classification Polyclonal
Concentration 1.5mg/mL
Conjugate Rhodamine
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, 0.01% Thimerosal and 0.05% Sodium Azide. RECONSTITUTION: Reconstitute to 1mg/mL with 1.5mL sterile distilled water.
Host Species Rabbit
Quantity 1.5 mg
Primary or Secondary Secondary
Target Species Goat
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only