missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Goat IgG (Heavy Chain), Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ 647, Invitrogen™

Catalog No. A27018
Encompass
Catalog No. A27018 Supplier Thermo Fisher Scientific Supplier No. LSA27018

Rabbit Recombinant Polyclonal Secondary Antibody

The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig, and donkey IgG is observed. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds – the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody – recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets – a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Goat IgG (Heavy Chain)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation PBS with 5mM sodium azide
Host Species Rabbit
Immunogen Recombinant full-length protein
Purification Method Gravity column chromatography
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Form Liquid
Isotype IgG
Show More Show Less