Learn More
Invitrogen™ Rabbit anti-Goat IgG (H+L) Secondary Antibody, AP
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31300 has been successfully used in Western blot applications. Product # 31300 reacts with the heavy chains of goat IgG and with the light chains common to most goat immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.0 mL of distilled water (0.6 mg/mL after restoration). Country of Origin: USA.
Specifications
Specifications
| Antigen | Goat IgG (H+L) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Alkaline Phosphatase (AP) |
| Formulation | 0.01M tris HCl with 0.25M NaCl, 15mg/mL BSA and 0.05% sodium azide; pH 8.0 |
| Host Species | Rabbit |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |