missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594
Description
Product will be shipped at Room Temperature.
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spécifications
Spécifications
| Antigène | Goat IgG (H+L) Cross-Adsorbed |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugué | Alexa Fluor 594 |
| Formule | PBS with 5mM sodium azide; pH 7.5 |
| Espèces hôtes | Rabbit |
| Immunogène | Gamma Immunoglobins Heavy and Light chains. |
| Méthode de purification | Purified |
| Quantité | 1 mg |
| Afficher plus de résultats |