missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG F(ab) Secondary Antibody [Janelia Fluor 525], R&D Systems™
Description
IgG F(ab) Polyclonal antibody specifically detects IgG F(ab) in Goat samples. It is validated for ELISA
Specifications
Specifications
| Antigen | IgG F(ab) |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Normal goat IgG |
| Purification Method | Affinity purified |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |