missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Goat, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ106FMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ106FMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ106FMI Supplier MilliporeSigma Supplier No. AQ106F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Rabbit anti-Goat IgG Antibody, F(ab′) 2, FITC conjugate detects level of Goat IgG and has been published and validated for use in immunofluoresence.

Specifications

Antigen Rabbit anti-Goat IgG F(ab')2
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Host Species Rabbit
Quantity 500 μg
Primary or Secondary Secondary
Target Species Goat

For Research Use Only