missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Goat, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP106CMI
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP106CMI 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP106CMI Supplier MilliporeSigma Supplier No. AP106C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

This Rabbit anti-Goat IgG Antibody, Cy3 conjugate is validated for use in IF for the detection of Goat IgG.

Specifications

Antigen Rabbit anti-Goat IgG
Applications Immunofluorescence
Classification Polyclonal
Conjugate Cyanine3
Host Species Rabbit
Quantity 1.5 mg
Primary or Secondary Secondary
Target Species Goat

For Research Use Only