missing translation for 'onlineSavingsMsg'
Learn More

IgY, bridging Rabbit anti-Chicken, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 12334MI
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12334MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12334MI Supplier MilliporeSigma Supplier No. 12334

Rabbit Polyclonal Antibody

Rabbit anti-Chicken IgY Antibody, bridging is an antibody against Chicken IgY for use in immunoprecipitation and western blotting.

Specifications

Antigen Rabbit anti-Chicken IgY
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M sodium phosphate, pH 7.4, 0.14M sodium chloride
Host Species Rabbit
Quantity 250 μg
Primary or Secondary Secondary
Target Species Chicken
Content And Storage −20°C for up to 24 months.
Form Powder
Show More Show Less

For Research Use Only