missing translation for 'onlineSavingsMsg'
Learn More

IgY Rabbit anti-Chicken, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 12341MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12341MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12341MI Supplier MilliporeSigma Supplier No. 12341

Rabbit Polyclonal Antibody

Detect Chicken IgY using this Rabbit anti-Chicken IgY Antibody, HRP conjugate validated for use in ELISA, western blot, immunohistochemistry.

Specifications

Antigen Rabbit anti-Chicken IgY
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation 0.02M Potassium Phosphate, 0.15M NaCl, pH 7.2, 10mg/mL BSA, and 0.01% gentamicin sulfate
Host Species Rabbit
Quantity 500 μg
Primary or Secondary Secondary
Target Species Chicken
Content And Storage 4°C for up to 24 months.

For Research Use Only