missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Chicken IgY (H+L) Secondary Antibody, FITC

Catalog No. PI31501
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31501 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31501 Supplier Invitrogen™ Supplier No. LSG31501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. This antibody has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Antibody Specificity: This antibody reacts with the heavy chains on chicken IgY and with the light chains common to most chicken immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins using immunoelectrophoresis. However, these antibodies may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 2-8°C until opened. Restore to 1.1 mL with distilled water (1.5 mg/mL after restoration). Centrifuge product if it is not clear upon standing for 1-2 hours at room temperature. To judge clarity, draw the product into a pasteur pipette. Product may be stored for several weeks at 2-8°C as undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorophore: Fluorescein-5-isothiocyanate (FITC-isomer 1) Amax = 492 nm; Emax = 520 nm Fluorophore/Protein: 8.5 μg/mg; 3.2 moles FITC per mole IgG Country of Origin: USA.

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Chicken IgY (H+L)
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate FITC
Formulation PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Host Species Rabbit
Purification Method Antigen affinity chromatography
Quantity 1.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG
Show More Show Less