missing translation for 'onlineSavingsMsg'
Learn More

Rabbit Anti-Chicken IgY Antibody (H&L) Antibody (HRP), ProSci

Catalog No. 16838639
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16838639 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16838639 Supplier ProSci Llc Supplier No. XG6183HRP0500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Control Antibody

Rabbit Anti-Chicken IgY Antibody specifically detects Rabbit Anti-Chicken IgY in Chicken samples. It is validated for ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen Rabbit Anti-Chicken IgY
Applications ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate HRP
Dilution 1:250-1:3,000 for IHC/ICC., , 1:2,000-1:5,000 for ELISA, Western Blot, color substrates., , 1:5,000-1:10,000 for Western Blot, chemiluminescence.
Formulation Buffer containing 0.01M sodium phosphate, 0.25M sodium chloride, pH 7.6 and 25% glycerol.
Host Species Rabbit
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity This antibody reacts with both heavy and light chains of chicken IgY, minimum cross-reactivity with IgG of mammalian species.
Target Species Chicken
Content And Storage Store 2 - 8 °C as an undiluted liquid for several weeks. After dilution, do not use for more than one day. -20 °C for up to one year.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less