missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Chicken, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP162FMI
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP162FMI 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP162FMI Supplier MilliporeSigma Supplier No. AP162F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Detect Chicken IgG using this Rabbit anti-Chicken IgG Antibody, FITC conjugate validated for use in immunofluoresence.

Specifications

Antigen Rabbit anti-Chicken IgG
Applications Immunofluorescence
Classification Polyclonal
Concentration 1.5mg/mL
Conjugate FITC
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6. Preservative 0.01% Thimerosal, 0.05% Sodium Azide with 15mg/mL BSA.
Host Species Rabbit
Quantity 1.5 mg
Primary or Secondary Secondary
Target Species Chicken
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only