missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | 1110046O21Rik; AGP1; AGPT; Agpt1; Ang1; Ang-1; angiopoietin 1; angiopoietin-1; ANGPT1; endothelial growth factor; KIAA0003 |
|---|---|
| Assay Range | 0.1 to 25 ng/mL |
| Assay Sensitivity | 0.1 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ER1RBX5
|
Invitrogen™
ER1RBX5 |
5 x 96 Tests |
N/A
|
|
|||||
ER1RB
|
Invitrogen™
ER1RB |
96 Tests |
N/A
|
|
|||||
Description
Rabbit Angiopoietin-1 quantitates rabbit Angiopoietin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rabbit Angiopoietin-1.
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.Specifications
| 1110046O21Rik; AGP1; AGPT; Agpt1; Ang1; Ang-1; angiopoietin 1; angiopoietin-1; ANGPT1; endothelial growth factor; KIAA0003 | |
| 0.1 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Angpt1 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Angiopoietin 1 | |
| 4 hrs 45 mins |
| 0.1 to 25 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 100342781 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |