missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB8A Recombinant Rabbit Monoclonal Antibody (23GB4900)
GREENER_CHOICE

Catalog No. MA552736
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552736 100 μL
MA552737 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552736 Supplier Invitrogen™ Supplier No. MA552736

Rabbit Recombinant Monoclonal Antibody

RAB8A Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The protein encoded by this gene is a member of the RAS superfamily which are small GTP/GDP-binding proteins with an average size of 200 amino acids. The RAS-related proteins of the RAB/YPT family may play a role in the transport of proteins from the endoplasmic reticulum to the Golgi and the plasma membrane. This protein shares 97%, 96%, and 51% similarity with the dog RAB8, mouse MEL, and mouse YPT1 proteins, respectively and contains the 4 GTP/GDP-binding sites that are present in all the RAS proteins. The putative effector-binding site of this protein is similar to that of the RAB/YPT proteins. However, this protein contains a C-terminal CAAX motif that is characteristic of many RAS superfamily members but which is not found in YPT1 and the majority of RAB proteins. Although this gene was isolated as a transforming gene from a melanoma cell line, no linkage between MEL and malignant melanoma has been demonstrable. This oncogene is located 800 kb distal to MY09B on chromosome 19p13.1.

Specifications

Antigen RAB8A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4900
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB8A
Gene Accession No. P35280, P55258, P61006
Gene Alias AA409338; cell line NK14 derived transforming oncogene; MEL; mel transforming oncogene (derived from cell line NK14); mel transforming oncogene (derived from cell line NK14)- RAB8 homolog; mel transforming oncogene (RAB8 homolog); oncogene c-mel; Rab8; Rab8a; RAB8A, member RAS oncogene family; ras-associated protein RAB8; ras-related protein Rab-8A
Gene Symbols RAB8A
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB8A (165-195AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117103, 17274, 4218
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less