missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB11FIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121227
Description
Positive test controls include: HeLa, HepG2, COS-7. The target is usually found in the following locations: Cytoplasmic vesicle, Recycling endosome, phagosome membrane. Immunogen sequence: EAKGEIKDSS PSSSPSPKGF RKKHLFSSTE NLAAGSWKEP AEGGGLSSDR QLSESSTKDS LKSMTLPSYR PAPLVSGDLR ENMAPANSEA TKEAKESKKP ESRRSSLLSL MTGKKDVAKG SEGENPLTVP GREKEGMLMG VKPGEDASGP AEDLVRRSEK DTAAVVSRQG SSLNLFEDVQ ITEPEAEPES KSEPRPPISS PRAPQTRAVK P.
Rab11-FIP1 (Rab 11 family-interacting protein 1), also known as Rab-coupling protein (RCP), is a 1283 amino acid Rab 11 effector protein. Rab11-FIP1, by interacting with Rab GTPases, is involved in the endosomal recycling process and may play a role in controlling membrane trafficking along the phagocytic pathway and during phagocytosis. Localized to the recycling endosome, the cytoplasmic membrane and phagosome membranes, Rab11-FIP1 is expressed as five isoforms produced by alternative splicing. As the most highly expressed isoform, isoform two of Rab11-FIP1 is expressed in brain, lung, testis, small intestine, spleen and heart. Isoform two of Rab11-FIP1 also has been found to form a homooligomer and is believed to interact with many Rab GTPases, including Rab 4A, Rab 11A, Rab 11B and Rab 25.Specifications
| RAB11FIP1 | |
| Polyclonal | |
| Unconjugated | |
| RAB11FIP1 | |
| 2010200K21Rik; 4833414G05Rik; CBBM; CBP; hCG_41347; NOEL1A; Rab coupling protein; Rab effector protein; RAB11 coupling protein; RAB11 family interacting protein 1; RAB11 family interacting protein 1 (class I); rab11 family-interacting protein 1; RAB11FIP1; Rab11-FIP1; rab11fip1 {ECO:0000250; rab-coupling protein; Rab-interacting recycling protein; Rcp; RGD1562965; UniProtKB:Q6WKZ4} | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 80223 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.72 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6WKZ4 | |
| RAB11FIP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 330-540 of human RAB11FIP1 (NP_079427.4). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |