missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB10 Monoclonal Antibody (4E2)
GREENER_CHOICE

Catalog No. PIMA515670
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515670 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515670 Supplier Invitrogen™ Supplier No. MA515670
Only null left

Mouse Monoclonal Antibody

MA5-15670 targets Rab10 in indirect ELISA, IF and WB applications and shows reactivity with Human and mouse samples. The MA5-15670 immunogen is purified recombinant fragment of human Rab10 expressed in E. Coli. . MA5-15670 detects Rab10 which has a predicted molecular weight of approximately 22.5kDa.

RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.

Specifications

Antigen RAB10
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4E2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene RAB10
Gene Accession No. P61026, P61027
Gene Alias Ac1075; AW107754; GTP-binding protein RAB10; Rab10; RAB10, member RAS oncogene family; ras-related GTP-binding protein; ras-related protein rab10; ras-related protein Rab-10
Gene Symbols RAB10
Host Species Mouse
Immunogen Purified recombinant fragment of human Rab10 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10890, 19325
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less