missing translation for 'onlineSavingsMsg'
Learn More

(R)-Styrene Oxide 96.0+%, TCI America™

Catalog No. p-7135915
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
S05165G 5 g
S051625G 25 g
2 options

Catalog No. S05165G

Supplier: TCI America S05165G

Only null left

Chemical Identifiers

CAS 20780-53-4
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00066210
InChI Key AWMVMTVKBNGEAK-QMMMGPOBSA-N
Synonym r-styrene oxide, r-phenyloxirane, r-+-styrene oxide, r-2-phenyloxirane, 2r-2-phenyloxirane, r-epoxyethyl benzene, oxirane, phenyl-, 2r, benzene, epoxyethyl-, r
PubChem CID 114705
ChEBI CHEBI:45389
IUPAC Name (2R)-2-phenyloxirane
SMILES C1O[C@@H]1C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 194°C
Quantity 5 g
UN Number 2810
Formula Weight 120.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-Styrene Oxide
EINECSNumber (3)-1033
RTECSNumber CZ9625010
TSCA No