missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Styrene oxide, 98%

Catalog No. AAB2124514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2124514 25 g
AAB2124506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2124514 Supplier Thermo Scientific Chemicals Supplier No. B2124514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20780-53-4 | C8H8O | 120.15 g/mol

It is used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine. It is the main metabolite of styrene, catalyzed by epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine. It is the main metabolite of styrene, catalyzed by epoxide hydrolase.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 20780-53-4
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00066210
InChI Key AWMVMTVKBNGEAK-QMMMGPOBSA-N
Synonym r-styrene oxide, r-phenyloxirane, r-+-styrene oxide, r-2-phenyloxirane, 2r-2-phenyloxirane, r-epoxyethyl benzene, oxirane, phenyl-, 2r, benzene, epoxyethyl-, r
PubChem CID 114705
ChEBI CHEBI:45389
IUPAC Name (2R)-2-phenyloxirane
SMILES C1O[C@@H]1C1=CC=CC=C1

Specifications

Density 1.051
Boiling Point 194°C
Flash Point 74°C (165°F)
Refractive Index 1.535
Quantity 25 g
UN Number UN2810
Beilstein 1984
Solubility Information Insoluble in water.
Optical Rotation +32° (Neat)
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material (R)-(+)-Styrene oxide
Show More Show Less
Hazard Category H227-H312-H315-H317-H319-H331-H340-H350
Hazard Statement GHS H Statement
H350-H312-H319-H227
May cause cancer.
Harmful in contact with skin.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECSNumber CZ9625010
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only