missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-SEGPHOS(regR) 99.0+%, TCI America™
Supplier: TCI America S09301G
Specifications
| (R)-(+)-SEGPHOS(regR) | |
| 172°C | |
| MFCD09753005 | |
| (R)-(+)-5,5′C-Bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole, (R)-(+)-4,4′C-Bis(diphenylphosphino)-3,3′C-bi(1,2-methylenedioxybenzene) | |
| C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 610.59 | |
| 610.59 | |
| Crystalline Powder |
| 244261-66-3 | |
| C38H28O4P2 | |
| 1 g | |
| RZZDRSHFIVOQAF-UHFFFAOYSA-N | |
| {4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane | |
| 11017510 | |
| ≥99.0% (HPLC) |
Chemical Identifiers
| 244261-66-3 | |
| 610.59 | |
| RZZDRSHFIVOQAF-UHFFFAOYSA-N | |
| 11017510 | |
| C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C38H28O4P2 | |
| MFCD09753005 | |
| (R)-(+)-5,5′C-Bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole, (R)-(+)-4,4′C-Bis(diphenylphosphino)-3,3′C-bi(1,2-methylenedioxybenzene) | |
| {4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane |
Safety and Handling
TSCA : No