missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-SEGPHOS(regR) 99.0+%, TCI America™
$98.79 - $337.40
Chemical Identifiers
| CAS | 244261-66-3 |
|---|---|
| Molecular Formula | C38H28O4P2 |
| Molecular Weight (g/mol) | 610.59 |
| MDL Number | MFCD09753005 |
| InChI Key | RZZDRSHFIVOQAF-UHFFFAOYSA-N |
| Synonym | (R)-(+)-5,5′C-Bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole, (R)-(+)-4,4′C-Bis(diphenylphosphino)-3,3′C-bi(1,2-methylenedioxybenzene) |
| PubChem CID | 11017510 |
| IUPAC Name | {4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane |
| SMILES | C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
S0930200MG
|
TCI America
S0930200MG |
200 mg |
Each for $98.79
|
|
|||||
|
S09301G
|
TCI America
S09301G |
1 g |
Each for $337.40
|
|
|||||
Chemical Identifiers
| 244261-66-3 | |
| 610.59 | |
| RZZDRSHFIVOQAF-UHFFFAOYSA-N | |
| 11017510 | |
| C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C38H28O4P2 | |
| MFCD09753005 | |
| (R)-(+)-5,5′C-Bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole, (R)-(+)-4,4′C-Bis(diphenylphosphino)-3,3′C-bi(1,2-methylenedioxybenzene) | |
| {4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane |
Specifications
| 244261-66-3 | |
| C38H28O4P2 | |
| 200 mg | |
| RZZDRSHFIVOQAF-UHFFFAOYSA-N | |
| {4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane | |
| 11017510 | |
| ≥99.0% (HPLC) | |
| (R)-(+)-SEGPHOS(regR) |
| 172°C | |
| MFCD09753005 | |
| (R)-(+)-5,5′C-Bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole, (R)-(+)-4,4′C-Bis(diphenylphosphino)-3,3′C-bi(1,2-methylenedioxybenzene) | |
| C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 610.59 | |
| 610.59 | |
| Crystalline Powder |
Safety and Handling
TSCA : No