missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-sec-Butanol, 99%

CAS: 14898-79-4 | C4H10O | 74.123 g/mol

Supplier:  Thermo Scientific Chemicals 217230050

 View more versions of this product

Catalog No. AC217230050

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(R)-(-)-sec-Butanol
Colorless
100.0
Authentic
Liquid
99%
C2H5CH(OH)CH3
01, I, 189
r---2-butanol, r-2-butanol, --2-butanol, 2r-butan-2-ol, r-butan-2-ol, r---sec-butanol, 2-butanol, 2r, unii-dlh38k423j, r---sec-butyl alcohol
CCC(C)O
74.123
CHEBI:35686
≥98.5%
14898-79-4
98.5
5 g
Glass Bottle
1.3960 to 1.3980 (20°C, 589nm)
C4H10O
MFCD00064280
15, 1543
BTANRVKWQNVYAZ-SCSAIBSYSA-N
(2R)-butan-2-ol
84682
74.12

Chemical Identifiers

14898-79-4
74.123
BTANRVKWQNVYAZ-SCSAIBSYSA-N
84682
(2R)-butan-2-ol
C4H10O
MFCD00064280
r---2-butanol, r-2-butanol, --2-butanol, 2r-butan-2-ol, r-butan-2-ol, r---sec-butanol, 2-butanol, 2r, unii-dlh38k423j, r---sec-butyl alcohol
CHEBI:35686
CCC(C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only