missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-sec-Butanol, 99%

Catalog No. AC217230010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217230010 1 g Glass Bottle
AC217230050 5 g Glass Bottle
AC217230250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC217230010 Supplier Thermo Scientific Chemicals Supplier No. 217230010

CAS: 14898-79-4 | C4H10O | 74.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14898-79-4
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00064280
InChI Key BTANRVKWQNVYAZ-SCSAIBSYSA-N
Synonym r---2-butanol, r-2-butanol, --2-butanol, 2r-butan-2-ol, r-butan-2-ol, r---sec-butanol, 2-butanol, 2r, unii-dlh38k423j, r---sec-butyl alcohol
PubChem CID 84682
ChEBI CHEBI:35686
IUPAC Name (2R)-butan-2-ol
SMILES CCC(C)O

Specifications

Color Colorless
CAS Min % 98.5
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula C2H5CH(OH)CH3
Beilstein 01, I, 189
Merck Index 15, 1543
Formula Weight 74.12
Percent Purity ≥98.5%
Chemical Name or Material (R)-(-)-sec-Butanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only