missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Rolipram 98.0+%, TCI America™

Catalog No. p-7135746
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
R018210MG 10 mg
R018250MG 50 mg
2 options

Catalog No. R018210MG

Supplier: TCI America R018210MG

Only null left

Chemical Identifiers

CAS 85416-75-7
Molecular Formula C16H21NO3
Molecular Weight (g/mol) 275.348
MDL Number MFCD03093861
InChI Key HJORMJIFDVBMOB-LBPRGKRZSA-N
Synonym r---rolipram, r-rolipram, r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, --rolipram, 4r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, unii-dpx51kup08, dpx51kup08, 1xmy, rolipram, r
PubChem CID 448055
ChEBI CHEBI:40133
IUPAC Name (4R)-4-(3-cyclopentyloxy-4-methoxyphenyl)pyrrolidin-2-one
SMILES COC1=C(C=C(C=C1)C2CC(=O)NC2)OC3CCCC3

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 10 mg
Formula Weight 275.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-Rolipram
TSCA No
Recommended Storage Freezer