missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Rolipram 98.0+%, TCI America™

Catalog No. R018210MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R018210MG 10 mg
R018250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R018210MG Supplier TCI America Supplier No. R018210MG
Only null left

Chemical Identifiers

CAS 85416-75-7
Molecular Formula C16H21NO3
Molecular Weight (g/mol) 275.348
MDL Number MFCD03093861
InChI Key HJORMJIFDVBMOB-LBPRGKRZSA-N
Synonym r---rolipram, r-rolipram, r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, --rolipram, 4r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, unii-dpx51kup08, dpx51kup08, 1xmy, rolipram, r
PubChem CID 448055
ChEBI CHEBI:40133
IUPAC Name (4R)-4-(3-cyclopentyloxy-4-methoxyphenyl)pyrrolidin-2-one
SMILES COC1=C(C=C(C=C1)C2CC(=O)NC2)OC3CCCC3

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 10 mg
Formula Weight 275.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-Rolipram
TSCA No
Recommended Storage Freezer