missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-N,N'-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine 98.0+%, TCI America™

Catalog No. d25201g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25201G Supplier TCI America Supplier No. D25201G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121788-73-6
Molecular Formula C16H14F6N2O4S2
Molecular Weight (g/mol) 476.408
MDL Number MFCD01321158
InChI Key XQAIGOHPAZPGOU-ZIAGYGMSSA-N
Synonym (R,R)-1,2-Bis(trifluoromethanesulfonamido)-1,2-diphenylethane
PubChem CID 2755245
IUPAC Name N-[(1R,2R)-1,2-diphenyl-2-(trifluoromethylsulfonylamino)ethyl]-1,1,1-trifluoromethanesulfonamide
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)NS(=O)(=O)C(F)(F)F)NS(=O)(=O)C(F)(F)F

Specifications

Melting Point 213°C
Color White
Quantity 1 g
Formula Weight 476.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-N,N′-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine