missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine 98.0+%, TCI America™
Supplier: TCI America B441825G
Specifications
| (R,R)-(-)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine | |
| 208°C | |
| C36H54N2O2 | |
| 25 g | |
| FGIQYENVFXNDTF-UHFFFAOYNA-N | |
| 2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}cyclohexyl)amino]methylidene}cyclohexa-2,4-dien-1-one | |
| 12014886 | |
| ≥98.0% (HPLC,N) |
| 135616-40-9 | |
| Yellow | |
| MFCD00191800 | |
| 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene, r,r---n,n'-bis 3,5-di-tert-butylsalicylidene-1,2-cyclohexanediamine, r,r-jacobsenliga, r,r-jacobsen ligand, 1r,2r---1,2-cyclohexane diamino-n,n'-bis 3,5-di-tert-butylsalicylidene, r,r---n,n'-bis 3,5-di-t-butylsalicylidene-1,2-cyclohexanediamine, 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene ,? r,r-jacobsen ligand | |
| CC(C)(C)C1=CC(=CNC2CCCCC2NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C | |
| 546.84 | |
| 546.84 | |
| Crystalline Powder |
Chemical Identifiers
| 135616-40-9 | |
| 546.84 | |
| FGIQYENVFXNDTF-UHFFFAOYNA-N | |
| 12014886 | |
| CC(C)(C)C1=CC(=CNC2CCCCC2NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C |
| C36H54N2O2 | |
| MFCD00191800 | |
| 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene, r,r---n,n'-bis 3,5-di-tert-butylsalicylidene-1,2-cyclohexanediamine, r,r-jacobsenliga, r,r-jacobsen ligand, 1r,2r---1,2-cyclohexane diamino-n,n'-bis 3,5-di-tert-butylsalicylidene, r,r---n,n'-bis 3,5-di-t-butylsalicylidene-1,2-cyclohexanediamine, 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene ,? r,r-jacobsen ligand | |
| 2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}cyclohexyl)amino]methylidene}cyclohexa-2,4-dien-1-one |
Safety and Handling
TSCA : No