missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine 98.0+%, TCI America™

Catalog No. p-7123526
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44185G 5 g
B441825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44185G Supplier TCI America Supplier No. B44185G

Chemical Identifiers

CAS 135616-40-9
Molecular Formula C36H54N2O2
Molecular Weight (g/mol) 546.84
MDL Number MFCD00191800
InChI Key FGIQYENVFXNDTF-UHFFFAOYNA-N
Synonym 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene, r,r---n,n'-bis 3,5-di-tert-butylsalicylidene-1,2-cyclohexanediamine, r,r-jacobsenliga, r,r-jacobsen ligand, 1r,2r---1,2-cyclohexane diamino-n,n'-bis 3,5-di-tert-butylsalicylidene, r,r---n,n'-bis 3,5-di-t-butylsalicylidene-1,2-cyclohexanediamine, 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene ,? r,r-jacobsen ligand
PubChem CID 12014886
IUPAC Name 2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}cyclohexyl)amino]methylidene}cyclohexa-2,4-dien-1-one
SMILES CC(C)(C)C1=CC(=CNC2CCCCC2NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C

Specifications

Melting Point 208°C
Color Yellow
Quantity 5 g
Formula Weight 546.84
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-(-)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine