missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R)-(+)-Hydrobenzoin 99.0+%, TCI America™
$149.13 - $497.70
Chemical Identifiers
| CAS | 52340-78-0 |
|---|---|
| Molecular Formula | C14H14O2 |
| Molecular Weight (g/mol) | 214.264 |
| MDL Number | MFCD00064254 |
| InChI Key | IHPDTPWNFBQHEB-ZIAGYGMSSA-N |
| Synonym | r,r-+-hydrobenzoin, 1r,2r-1,2-diphenylethane-1,2-diol, r,r-hydrobenzoin, +-hydrobenzoin, unii-wv0i33nond, +-1r,2r-1,2-diphenylethane-1,2-diol, wv0i33nond, isohydrobenzoin, 1,2-ethanediol, 1,2-diphenyl-, 1r,2r-rel, +-dihydrobenzoin |
| PubChem CID | 853019 |
| ChEBI | CHEBI:50014 |
| IUPAC Name | (1R,2R)-1,2-diphenylethane-1,2-diol |
| SMILES | C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O |
Chemical Identifiers
| 52340-78-0 | |
| 214.264 | |
| IHPDTPWNFBQHEB-ZIAGYGMSSA-N | |
| 853019 | |
| (1R,2R)-1,2-diphenylethane-1,2-diol |
| C14H14O2 | |
| MFCD00064254 | |
| r,r-+-hydrobenzoin, 1r,2r-1,2-diphenylethane-1,2-diol, r,r-hydrobenzoin, +-hydrobenzoin, unii-wv0i33nond, +-1r,2r-1,2-diphenylethane-1,2-diol, wv0i33nond, isohydrobenzoin, 1,2-ethanediol, 1,2-diphenyl-, 1r,2r-rel, +-dihydrobenzoin | |
| CHEBI:50014 | |
| C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O |
Specifications
| 52340-78-0 | |
| White | |
| MFCD00064254 | |
| r,r-+-hydrobenzoin, 1r,2r-1,2-diphenylethane-1,2-diol, r,r-hydrobenzoin, +-hydrobenzoin, unii-wv0i33nond, +-1r,2r-1,2-diphenylethane-1,2-diol, wv0i33nond, isohydrobenzoin, 1,2-ethanediol, 1,2-diphenyl-, 1r,2r-rel, +-dihydrobenzoin | |
| C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O | |
| 214.264 | |
| CHEBI:50014 | |
| ≥99.0% (GC) | |
| (R,R)-(+)-Hydrobenzoin |
| 148°C | |
| C14H14O2 | |
| 5 g | |
| IHPDTPWNFBQHEB-ZIAGYGMSSA-N | |
| (1R,2R)-1,2-diphenylethane-1,2-diol | |
| 853019 | |
| 214.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No