CAS RN 52340-78-0
IUPAC Name:
(1R,2R)-1,2-diphenylethane-1,2-diol
Synonyms:
(+)-hydrobenzoin (R,R)-hydrobenzoin (R,R)-(+)-hydrobenzoin
Molecular Weight (g/mol):
214.26
Molecular Formula:
C14H14O2
InChi Key:
IHPDTPWNFBQHEB-ZIAGYGMSSA-N
SMILES:
O[C@@H]([C@H](O)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(R,R)-(+)-Hydrobenzoin 99.0+%, TCI America™
CAS: 52340-78-0 Molecular Formula: C14H14O2 Molecular Weight (g/mol): 214.264 MDL Number: MFCD00064254 InChI Key: IHPDTPWNFBQHEB-ZIAGYGMSSA-N Synonym: r,r-+-hydrobenzoin,1r,2r-1,2-diphenylethane-1,2-diol,r,r-hydrobenzoin,+-hydrobenzoin,unii-wv0i33nond,+-1r,2r-1,2-diphenylethane-1,2-diol,wv0i33nond,isohydrobenzoin,1,2-ethanediol, 1,2-diphenyl-, 1r,2r-rel,+-dihydrobenzoin PubChem CID: 853019 ChEBI: CHEBI:50014 IUPAC Name: (1R,2R)-1,2-diphenylethane-1,2-diol SMILES: C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O