missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(+)-Hydrobenzoin 99.0+%, TCI America™

Catalog No. H08185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08185G 5 g
H081825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08185G Supplier TCI America Supplier No. H08185G
Only null left

Chemical Identifiers

CAS 52340-78-0
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00064254
InChI Key IHPDTPWNFBQHEB-ZIAGYGMSSA-N
Synonym r,r-+-hydrobenzoin, 1r,2r-1,2-diphenylethane-1,2-diol, r,r-hydrobenzoin, +-hydrobenzoin, unii-wv0i33nond, +-1r,2r-1,2-diphenylethane-1,2-diol, wv0i33nond, isohydrobenzoin, 1,2-ethanediol, 1,2-diphenyl-, 1r,2r-rel, +-dihydrobenzoin
PubChem CID 853019
ChEBI CHEBI:50014
IUPAC Name (1R,2R)-1,2-diphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O

Specifications

Melting Point 148°C
Color White
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-(+)-Hydrobenzoin