missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(-)-2,3-Butanediol 97.0+%, TCI America™

Catalog No. B11611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11611G 1 g
B11615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11611G Supplier TCI America Supplier No. B11611G

Chemical Identifiers

CAS 24347-58-8
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00064267
InChI Key OWBTYPJTUOEWEK-UHFFFAOYNA-N
Synonym 2r,3r-butane-2,3-diol, r,r-2,3-butanediol, 2r,3r---2,3-butanediol, r,r-butane-2,3-diol, 2r,3r-2,3-butanediol, r,r-2,3-butylene glycol, r,r---butane-2,3-diol, r,r---2,3-butylene glycol, unii-or02b2286a, r,r---2,3-butanediol
PubChem CID 225936
ChEBI CHEBI:16982
IUPAC Name butane-2,3-diol
SMILES CC(O)C(C)O

Specifications

Color Yellow
Boiling Point 80°C
Quantity 1 g
Formula Weight 90.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (R,R)-(-)-2,3-Butanediol
EINECSNumber (2)-0235
TSCA No