accessibility menu, dialog, popup

UserName

H3COHCH3HO

CAS RN 24347-58-8

IUPAC Name: butane-2,3-diol
Synonyms: 2,3-butanediol dimethylethylene glycol dimethylene glycol 2,3-butanediol (meso) pseudobutylene glycol sym-dimethylethylene glycol 2,3-butylene glycol
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: OWBTYPJTUOEWEK-UHFFFAOYNA-N
SMILES: CC(O)C(C)O
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)