missing translation for 'onlineSavingsMsg'
Learn More

(R,R″)-2,2″-Bis(diphenylphosphino)-1,1″-biferrocene 98.0+%, TCI America™

Catalog No. B3196100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3196100MG 100 mg
B3196500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3196100MG Supplier TCI America Supplier No. B3196100MG

Chemical Identifiers

CAS 136274-57-2
Molecular Formula C44H36Fe2P2
Molecular Weight (g/mol) 738.41
InChI Key VADPYXYBFVOWCG-UHFFFAOYSA-N
PubChem CID 44629959
IUPAC Name cyclopenta-1,3-diene;[2-(2-diphenylphosphanylcyclopenta-1,3-dien-1-yl)cyclopenta-1,4-dien-1-yl]-diphenylphosphane;iron(2+)
SMILES [CH-]1C=CC=C1.[CH-]1C=CC=C1.[CH-]1C=CC(=C1C2=C(C=C[CH-]2)P(C3=CC=CC=C3)C4=CC=CC=C4)P(C5=CC=CC=C5)C6=CC=CC=C6.[Fe+2].[Fe+2]

Specifications

Melting Point 232°C
Quantity 100 mg
Formula Weight 738.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (R,R′′)-2,2′′-Bis(diphenylphosphino)-1,1′′-biferrocene