CAS RN 136274-57-2
IUPAC Name:
bis(λ²-iron(2+)) 5,5'-bis(diphenylphosphanyl)-[1,1'-bi(cyclopentane)]-1(5),1'(5'),3,3'-tetraene-2,2'-diide bis(cyclopenta-2,4-dien-1-ide)
Synonyms:
bis(λ²-iron(2+)) 5,5'-bis(diphenylphosphanyl)-[1,1'-bi(cyclopentane)]-1(5),1'(5'),3,3'-tetraene-2,2'-diide bis(cyclopentadienide)
Molecular Weight (g/mol):
738.41
Molecular Formula:
C44H36Fe2P2
InChi Key:
VADPYXYBFVOWCG-UHFFFAOYSA-N
SMILES:
[Fe++].[Fe++].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[CH-]1C=CC(P(C2=CC=CC=C2)C2=CC=CC=C2)=C1C1=C(C=C[CH-]1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(R,R″)-2,2″-Bis(diphenylphosphino)-1,1″-biferrocene 98.0+%, TCI America™
CAS: 136274-57-2 Molecular Formula: C44H36Fe2P2 Molecular Weight (g/mol): 738.41 InChI Key: VADPYXYBFVOWCG-UHFFFAOYSA-N PubChem CID: 44629959 IUPAC Name: cyclopenta-1,3-diene;[2-(2-diphenylphosphanylcyclopenta-1,3-dien-1-yl)cyclopenta-1,4-dien-1-yl]-diphenylphosphane;iron(2+) SMILES: [CH-]1C=CC=C1.[CH-]1C=CC=C1.[CH-]1C=CC(=C1C2=C(C=C[CH-]2)P(C3=CC=CC=C3)C4=CC=CC=C4)P(C5=CC=CC=C5)C6=CC=CC=C6.[Fe+2].[Fe+2]