missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Propylene Oxide 98.0+%, TCI America™

Catalog No. P13965ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13965ML 5 mL
P139625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13965ML Supplier TCI America Supplier No. P13965ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15448-47-2
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00066211
InChI Key GOOHAUXETOMSMM-GSVOUGTGSA-N
Synonym r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane
PubChem CID 146261
ChEBI CHEBI:28985
IUPAC Name (2R)-2-methyloxirane
SMILES CC1CO1

Specifications

Color Colorless
Boiling Point 34°C
Quantity 5 mL
UN Number 1280
Formula Weight 58.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-Propylene Oxide
EINECSNumber (2)-0219
RTECSNumber UJ2650000
TSCA No
Recommended Storage Refrigerator