missing translation for 'onlineSavingsMsg'
Learn More

(R)-Phenylephrine Hydrochloride 98.0+%, TCI America™

Catalog No. P03985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03985G 5 g
P039825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03985G Supplier TCI America Supplier No. P03985G

Chemical Identifiers

CAS 61-76-7
Molecular Formula C9H14ClNO2
Molecular Weight (g/mol) 203.67
MDL Number MFCD00012605,MFCD00044749
InChI Key OCYSGIYOVXAGKQ-UHFFFAOYNA-N
Synonym phenylephrine hydrochloride, phenylephrine hcl, l-phenylephrine hydrochloride, metaoxedrine chloride, neosympatol, synethenate, almefrin, consdrin, emagrin, fenilfar
PubChem CID 5284443
ChEBI CHEBI:8094
IUPAC Name hydrogen 3-[1-hydroxy-2-(methylamino)ethyl]phenol chloride
SMILES [H+].[Cl-].CNCC(O)C1=CC=CC(O)=C1

Specifications

Melting Point 143°C
Color White
Quantity 5 g
Formula Weight 203.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-Phenylephrine Hydrochloride
RTECSNumber DO7525000
TSCA Yes