missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(p-Toluenesulfinyl)ferrocene 98.0+%, TCI America™
$745.62 - $2399.03
Chemical Identifiers
| CAS | 130225-27-3 |
|---|---|
| Molecular Formula | C17H16FeOS |
| Synonym | (R)-Ferrocenyl p-Tolyl Sulfoxide |
Chemical Identifiers
| 130225-27-3 | |
| C17H16FeOS | |
| (R)-Ferrocenyl p-Tolyl Sulfoxide |
Specifications
| 130225-27-3 | |
| C17H16FeOS | |
| (R)-Ferrocenyl p-Tolyl Sulfoxide | |
| ≥98.0% (HPLC,T) | |
| (R)-(p-Toluenesulfinyl)ferrocene |
| 143°C | |
| 1 g | |
| 324.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No