accessibility menu, dialog, popup

UserName

Découvrez nos nouvelles offres vedettes sur l’équipement, les instruments et plus encore

Économisez maintenant

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 130225-27-3

A(a)

CAS RN 130225-27-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.