missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-Naproxen Pharmaceutical Impurity Standard, ≥98% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 23979-41-1 |
| Linear Formula | CH3 OC10H6 CH(CH3)CO2 H |
| MDL Number | MFCD00870716 |
| Melting Point | 156°C to 158°C |
| Synonym | (R)-(-)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid; Naproxen Impurity G; (2 R)-2-(6-Methoxy-2-naphthyl)propionic acid; (2 R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid; (R)-Naproxen |
| Type | Pharmaceutical Impurity Standard |
| Quantity | 50 mg |
| Formula Weight | 230.26 |
| Percent Purity | ≥98% (HPLC) |