missing translation for 'onlineSavingsMsg'
Learn More

(R)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine 96.0+%, TCI America™

Catalog No. d2535100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2535100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2535100MG Supplier TCI America Supplier No. D2535100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55700-44-2
Molecular Formula C26H28FeNP
Molecular Weight (g/mol) 441.34
MDL Number MFCD00075286
InChI Key RCAFPSZHKFOLEE-KVEBOLLKNA-N
Synonym (R)-(S)-PPFA
IUPAC Name (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine
SMILES [Fe].c1cccc1.C[C@@H](N(C)C)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 100 mg
Formula Weight 441.34
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine